주요 문서
BAH942404002
Scienceware® Lab Companion 3-way valve
fits all cabinet vacuum desiccators, pack of 1 ea
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104510
특징
connects to 1/4 in. tubing
포장
pack of 1 ea
제조업체/상표
Bel-Art H94240-4002
일반 설명
Bel-Art Viton 3-Way Valve, available as a replacement of or addition to Lab Companion cabinet-style vacuum desiccators.
특징 및 장점
  • Viton® 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange
  • Four built-in ports are standard on the cabinet style units and one Viton® 3-way valve is included; attachment of up to three additional valves is possible
법적 정보
Lab Companion is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)