주요 문서
BAH186101440
Stak-A-Tray large tray
W × L 35.6 cm × 35.6 cm
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122800
NACRES:
NB.35
포장
pack of 1 ea
제조업체/상표
Bel-Art H18610-1440
너비 × 길이
35.6 cm × 35.6 cm
일반 설명
Modular rack and tray system allows multi-layer storage and prevents tipping of stacked flasks that result in spills.
특징 및 장점
  • Can be used with Petri dishes and cell culture flasks in incubators, cold rooms or on the bench
  • Each tier accommodates one large or two small trays
  • Stack up to five racks for increased storage within the same footprint
  • Perforated trays allow air flow and slide easily in the rack
  • All parts are stainless steel; autoclavable at 121ºC (250ºF)
  • Racks and trays sold separately
법적 정보
Stak-A-Tray is a trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)