주요 문서
BAF249660008
SP Bel-Art® Splash Shield Weighted base Mount
round, diam. 9.5" (24.1 cm)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122800
NACRES:
NB.43
material
round
포장
pack of 1 ea
제조업체/상표
Bel-Art F24966-0008
직경
9.5" (24.1 cm)
일반 설명
No matter how your workspace is configured, these Plexiglas® Splash Shields are designed to provide safety plus comfort (not hot and stuffy like personal face shields), flexibility (they’re fully adjustable), and quality in a design that will adapt to your specific needs.
특징 및 장점
  • All shields move side-to-side as well as up and down at the touch of a finger
  • Free precious work space and enhance the performance of every user
  • After selecting desired shield, order appropriate mounting fixtures to suit your work area
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Plexiglas is a registered trademark of Arkema
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)