주요 문서
BAF170720010
Cleanware microscope optics, additional optical cleaning pads
pack of 10 ea
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103200
NACRES:
NB.35
포장
pack of 10 ea
제조업체/상표
Bel-Art F17072-0010
일반 설명
Contains everything you need to keep optics and microscope parts sanitary, clean and clear. Ten pre-saturated optical cleaning pads, two individually wrapped packs of anti-static plastic surface wipes and Two, 15.2cm (6") swabs pre-moistened with alcohol, contains complete instructions for use.
법적 정보
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)