주요 문서
BAF126280000
Pouring spout with fingertip control
pack of 12 ea, high-density polyethylene
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41100000
NACRES:
NB.51
material
high-density polyethylene
포장
pack of 12 ea
제조업체/상표
Bel-Art F12628-0000
일반 설명
Chemical resistant, high-density polyethylene spout has a small air hole that allows finger tip control of pouring speed.
  • Fits 38 mm tall buttress closure bottles
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)