주요 문서
BAB615065100
SP Bel-Art® Durac® Bluetooth Thermometer / Hygrometer with 30 day logger
temp. range at -10-50 °C, humidity limit 0-99%
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41112303
NACRES:
NB.79
포장
pack of 1 ea
제조업체/상표
HB Instruments B61506-5100
파라미터
temp. range at -10-50 °C
0-99% humidity limit
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Enable remote monitoring and alerts by broadcasting readings to any Apple or Android smartphone or device with Bluetooth 4.0 or better utilizing the free THERMSmart app. Available in verification and environmental monitoring models.
특징 및 장점
  • Monitor critical environments without a physical connection; maximum signal distance is approximately 40 meters
  • Record temperature and humidity data for 30 days
  • Hourly data recorded for current seven days, then maximum and minimum temperatures and humidity are recorded for next 23 days
  • Verification model records accurate temperature reading without sample contamination by using a glass bead filled bottle (included) which mimics the sample
  • Alarm sounds to alert user if temperature falls outside of the set parameters
  • Data viewable as a table, graph, or calendar; data from up to six data loggers can be plotted on one graph for direct comparison; also downloadable as a CSV file
  • Indicators include memory full, active alarm state and low battery; Hygrometer WBGT (Wet Bulb Global Temperature), Heat, Mold and Comfort indices
  • App can trigger indicator light for unit identification
  • Includes Certificate of Compliance indicating accuracy for 4 temperature and 3 humidity points, and multi-language (English, French, German, Italian, Portuguese and Spanish) instructions
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Durac is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)