ZRB3203
Anti-CXCL1/GROα Antibody, clone 2J19 ZooMAb® Rabbit Monoclonal


recombinant, expressed in HEK 293 cells
Synonym(s):
Growth-regulated alpha protein
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2J19, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 11.3 kDa
observed mol wt ~11 kDa
purified by
using Protein A
species reactivity
human
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... CXCL1(2919)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in BT-549 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CXCL1/GRO in BT-549 cell lysate.
Tested Applications
Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected CXCL1/GRO in lysates from PC-3 cells and human spleen tissue.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected His-tagged, full-length, recombinant human CXCL1/GRO .
Affinity Binding Assay: A representative lot of this antibody bound with CXCL1/GRO peptide with at least ten thousand-fold (10,000X) higher than with non-specific control peptide.
Immunocytochemistry Analysis: A 1:100 Dilution from a representative lot detected CXCL1/GRO in PC-3 cells.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CXCL1/GRO in human spleen and human colon cancer tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service