Skip to Content
Merck
All Photos(11)

Key Documents

ZRB2745

Sigma-Aldrich

Anti-CXCL9/MIG Antibody, clone 1H13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

C-X-C motif chemokine 9;Gamma-interferon-induced monokine;Monokine induced by interferon-gamma;HuMIG;MIG;Small-inducible cytokine B9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1H13, recombinant monoclonal

description

1H13 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 14.02 kDa
observed mol wt ~38 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CXCL9(4283)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1H3 is a is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects CXCL9 protein.

Immunogen

GST/His-tagged Human recombinant mature CXCL9 (without the signal peptide).

Application

Quality Control Testing

Evaluated by Western Blotting with GST/His-tagged recombinant human CXCL9.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected recombinant Human CXCL9.

Tested applications

Affinity Binding Assay: A representative lot of this antibody bound recombinant CXCL9/MIG with a KD of 2.3 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CXCL9/MIG in THP-1 cells treated with IFN- and TNF- (both at 100 ng/mL; 24 h).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting with GST/His-tagged recombinant human CXCL9.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected recombinant Human CXCL9.

Target description

C-X-C motif chemokine 9 (UniProt: Q07325; also known as Gamma-interferon-induced monokine, Monokine induced by interferon-gamma, HuMIG, MIG, Small-inducible cytokine B9) is encoded by the CXCL9 (also known as CMK, MIG, SCYB9) gene (Gene ID: 4283) in human. CXCL9 is a small cytokine of the CXC chemokine family that lacks the Glu-Leu-Arg (ELR) domain. It is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off to generate the mature form. It is induced in macrophages in response to interferon- and is involved in growth, movement , and activation state of cells that participate in immune and inflammatory responses. It also acts as a chemotactic agent for activated T cells. Its actions are mediated via CXC-chemokine receptor 3 (CXCR3). Overexpression of CXCL9 is reported to reduced tumor progression and metastasis via the inhibition of angiogenesis. CXCL9 is reported to exert its tumor-suppressive function by recruiting tumor-infiltrating lymphocytes in human ovarian cancer. Conversely, CXCL9 can also act directly on tumor cells expressing the CXCR3 receptor to promote cell migration and epithelial mesenchymal transition. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Bronger, H., et al. (2016). Br. J. Cancer. 115(5); 553-563).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service