Skip to Content
Merck
All Photos(6)

Key Documents

ZRB2670

Anti-Mucolipin-1 Antibody, clone 1D9 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

MG-2, ML1, Mucolipidin, TRPML1, Transient receptor potential channel mucolipin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D9, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 65.02 kDa
observed mol wt ~70 kDa

purified by

using Protein A

species reactivity

human, mouse

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... MCOLN1(57192)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1D9 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects Mucolipin-1. It targets an epitope within 17 amino acids from the N-terminal half, cytoplasmic Domain.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal half, cytoplasmic Domain of human Mucolipin-1.

Application

Quality Control Testing

Evaluated by Western Blotting in mouse spleen tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Mucolipin-1 in mouse spleen tissue lysate.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Anti-Mucolipin-1 in human kidney tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Mucolipin-1 in U-2 OS cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Mucolipin-1 (UniProt:Q9GZU1; also known as ML1, MG-2, Mucolipidin, Transient receptor potential channel mucolipin 1, TRPML1) is encoded by the MCOLN1 (also known as ML4, TRPML1) gene (Gene ID: 57192) in human. Mucolipin-1 is a member of the small family of the TRPML channels, a subgroup of the large protein family of TRP ion channels. It is a multi-pass membrane protein and functions as a non-selective cation channel. It is widely expressed in adult and fetal tissues and mostly found in vesicles, and is involved in the regulation of lysosomal trafficking. The most N-terminal extracellular/lumenal domain (referred to as I-II linker or polycystin-mucolipin domain) contributes to a structure with a four-fold rotational symmetry in a tetrameric assembly. The protruding structure formed by the I-II linkers may contain all the interaction sites with lipids and proteins in the endo- lysosomal lumen. The structure contains a central highly electronegative pore with a 14 A diameter. The pore is critical for Ca2+ and pH regulation. Mucolipin-1 aids in the transport of Ca2+ into the cytosol from the lumen, in response to the changing levels of phosphatidylinositol-3, 5-bisphosphate. It is also required for efficient uptake of large particles in macrophages in which Ca2+ release from the lysosomes triggers lysosomal exocytosis. May also play a role in phagosome-lysosome fusion. Mutations in the MCOLN1 gene is implicated in Mucolipidosis type IV (MLIV) is an autosomal recessive, neurodegenerative disorder characterized by developmental abnormalities of the brain and impaired neurological, ophthalmologic and gastric function. MLIV pathophysiology has been linked to deficiency in membrane trafficking, and organelle dynamics in the late endocytic pathway. Specifically, MLIV cells have been shown to accumulate autophagosomes, due to increased de novo autophagosome formation and due to delayed fusion of autophagosomes with late endosomes/lysosomes. MCOLN1 might also be involved with Alzheimer s disease and HIV. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Li, M., et al. (2017). Nature Structural & Molecular Biology. 24:205-213; Raychowdhury M.K., et al. (2004). Hum. Mol. Genet. 13:617-627).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service