Skip to Content
Merck
All Photos(9)

Key Documents

ZRB2305

Sigma-Aldrich

Anti-MAP3K8 Antibody, clone 1D11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cancer Osaka thyroid oncogene, EC:2.7.11.25, Mitogen-activated protein kinase kinase kinase 8, Proto-oncogene c-Cot, Serine/threonine-protein kinase cot, TPL-2, Tumor progression locus 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D11, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 52.93 kDa
observed mol wt ~55 kDa

purified by

using Protein A

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1D11 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Mitogen-activated protein kinase kinase kinase 8 (MAP3K8). It targets an epitope within 17 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal half of human Mitogen-activated protein kinase kinase kinase 8 (MAP3K8).

Application

Quality Control Testing

Evaluated by Western Blotting in L6 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MAP3K8 in L6 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected MAP3K8 in lysate from A549 cells and 1:10,000 dilution in lysates from NIH3T3, and HeLa cells.

Affinity Binding Assay: A representative lot of this antibody bound MAP3K8 peptide with a KD of 1.8 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100-1,000 dilution from a representative lot detected MAP3K8 in Human brain, Rat brain and Human liver tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Mitogen-activated protein kinase kinase kinase 8 (UniProt: P41279; also known as EC:2.7.11.25, Cancer Osaka thyroid oncogene, Proto-oncogene c-Cot, Serine/threonine-protein kinase cot, Tumor progression locus 2, TPL-2) is encoded by the MAP3K8 (also known as COT, ESTF) gene (Gene ID: 1326) in human. MAP3K8 is involved in the regulation of MAPK cascades and immune responses. Its differential expression and dysregulation are closely correlated with tumorigenesis. It is expressed in several normal tissues and human tumor-derived cell lines. Two isoforms of MAP3K8 have been described that are produced by alternative splicing. It is shown to be involved in LPS-induced, TLR4-mediated activation of the MAPK/ERK pathway in macrophages. It is critical for production of the TNF-α during immune responses. It is also involved in the regulation of T-helper cell differentiation and IFN-γ expression in T-cells.  MAP3K8 undergoes autophosphorylation: isoform 1 undergoes phosphorylation mainly on serine residues whereas isoform 2 undergoes phosphorylation on both serine and threonine residues. Its phosphorylation on threonine 290 is necessary but is not sufficient for its full kinase activity. Its phosphorylation on serine 400 by IKK-β is require or LPS-stimulated activation of MAPK pathway in macrophages. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Hao, J., et al. (2021). Front. Genet. 12; 674613; Roget, K., et al. (2012). Mol. Cell Biol. 32(22); 4684-4690; Aoki, M., et al. (1993). J. Biol. Chem. 268(30); 22723-22132).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service