SRP0312
AEBP2 human
recombinant, expressed in baculovirus infected Sf9 cells, ≥48% (SDS-PAGE)
Synonym(s):
AE binding protein 2 (AEBP2)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥48% (SDS-PAGE)
form
aqueous solution
mol wt
53.6 kDa
packaging
pkg of 50 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... AEBP2(121536)
General description
Human AE binding protein 2 (AEBP2), (GenBank Accession No. NM_153207), amino acids 2-503 (end) with N-terminal His-tag, MW=53.6 kDa, expressed in Sf9 cells via a Baculovirus expression system.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hana Kim et al.
PloS one, 6(9), e25174-e25174 (2011-09-29)
Aebp2 is a potential targeting protein for the mammalian Polycomb Repression Complex 2 (PRC2). We generated a mutant mouse line disrupting the transcription of Aebp2 to investigate its in vivo roles. Aebp2-mutant homozygotes were embryonic lethal while heterozygotes survived to
Ru Cao et al.
Molecular cell, 15(1), 57-67 (2004-07-01)
Recent studies have revealed the intrinsic histone methyltransferase (HMTase) activity of the EED-EZH2 complex and its role in Hox gene silencing, X inactivation, and cancer metastasis. In this study, we focus on the function of individual components. We found that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service