Skip to Content
Merck
All Photos(1)

Key Documents

SBR00081

Sigma-Aldrich

Favipiravir

≥97% (HPLC), powder

Synonym(s):

Favilavir, T-705

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4FN3O2
CAS Number:
Molecular Weight:
157.10
UNSPSC Code:
51101500

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to yellow

antibiotic activity spectrum

viruses

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

C1=C(N=C(C(=O)N1)C(=O)N)F

InChI

InChI=1S/C5H4FN3O2/c6-2-1-8-5(11)3(9-2)4(7)10/h1H,(H2,7,10)(H,8,11)

InChI key

ZCGNOVWYSGBHAU-UHFFFAOYSA-N

General description

Favipiravir is a pyrazinecarboxamide derivative which is an RNA polymerase inhibitors. Favipiravir is converted to the ribofuranosyltriphosphate derivative by host enzymes and selectively inhibits the influenza viral RNA-dependent RNA polymerase. Favipiravir has broad-spectrum activity against RNA viruses, including influenza viruses, arena viruses, bunyaviruses, and flaviviruses.

Biochem/physiol Actions

Favipiravir inhibits the RNA-dependent RNA polymerase enzyme, which is essential for the replication of RNA viruses such as influenza and SARS-CoV-2 (the virus that causes COVID-19). Inhibition of RNA-dependent RNA polymerase activity leads to the disruption of viral RNA replication and ultimately, the reduction of viral load.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service