Skip to Content
Merck
All Photos(1)

Key Documents

SBR00080

Sigma-Aldrich

Dolutegravir

Synonym(s):

GSK 1349572

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19F2N3O5
CAS Number:
Molecular Weight:
419.38
UNSPSC Code:
51101500

Assay

≥98% (HPLC)

Quality Level

form

powder

color

white to off-white

antibiotic activity spectrum

viruses

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

C[C@@H]1CCO[C@@H]2N1C(=O)C3=C(C(=O)C(=CN3C2)C(=O)NCC4=C(C=C(C=C4)F)F)O

InChI

1S/C20H19F2N3O5/c1-10-4-5-30-15-9-24-8-13(17(26)18(27)16(24)20(29)25(10)15)19(28)23-7-11-2-3-12(21)6-14(11)22/h2-3,6,8,10,15,27H,4-5,7,9H2,1H3,(H,23,28)/t10-,15+/m1/s1

InChI key

RHWKPHLQXYSBKR-BMIGLBTASA-N

General description

Dolutegravir is an antibioitic which belongs to the class of compounds known as integrase strand transfer inhibitors (INSTIs).

Biochem/physiol Actions

Dolutegravir inhibits HIV-1 viral replication, high potency against the HIV-1 Y143R, N155H, and G140S/Q148H mutants. Dolutegravir inhibitis the action of the HIV integrase enzyme, which is necessary for the integration of viral DNA into the host genome, thereby preventing viral replication and reducing HIV viral load.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service