Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702692

Sigma-Aldrich

Anti-JARID2 Antibody, clone 3L5C5, Rabbit Monoclonal

Synonym(s):

JMJ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

3L5C5, monoclonal

form

liquid

mol wt

150 kDa

species reactivity

human, rat, rat

concentration

0.7mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

FEEGPSRKRPRLQAQRKFAQSQPNSPSTTPVKIVEPLLPPPATQISDLSKRKPKTEDFLTFLCLRGSPALPNSMVYFGSSQDEEEVEEEDDETEDVKTATN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JARID2(3720)

General description

This gene encodes a Jumonji- and AT-rich interaction domain (ARID)-domain-containing protein. The encoded protein is a DNA-binding protein that functions as a transcriptional repressor. This protein interacts with the Polycomb repressive complex 2 (PRC2) which plays an essential role in regulating gene expression during embryonic development. This protein facilitates the recruitment of the PRC2 complex to target genes. Alternate splicing results in multiple transcript variants. Mutations in this gene are associated with chronic myeloid malignancies. [provided by RefSeq, May 2012]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 100-200 of human JARID2 (Q92833).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service