Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702586

Sigma-Aldrich

Anti-ULK3 Antibody, clone 5Z7Z6, Rabbit Monoclonal

Synonym(s):

unc-51 like kinase 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

5Z7Z6, monoclonal

form

liquid

species reactivity

rat, rat, human

concentration

0.70 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

KPRLLAALEVASAAMAKEEAAGGEQDALDLYQHSLGELLLLLAAEPPGRRRELLHTEVQNLMARAEYLKEQVKMRESRWEADTLDKEGLSESVRSSCTLQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ULK3(25989)

General description

Serine/threonine protein kinase that acts as a regulator of Sonic hedgehog (SHH signaling and autophagy. Acts as a negative regulator of SHH signaling in the absence of SHH ligand: interacts with SUFU, thereby inactivating the protein kinase activity and preventing phosphorylation of GLI proteins (GLI1, GLI2 and/or GLI3. Positively regulates SHH signaling in the presence of SHH: dissociates from SUFU, autophosphorylates and mediates phosphorylation of GLI2, activating it and promoting its nuclear translocation. Phosphorylates in vitro GLI2, as well as GLI1 and GLI3, although less efficiently. Also acts as a regulator of autophagy: following cellular senescence, able to induce autophagy.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 373-472 of human ULK3 (Q6PHR2).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service