Skip to Content
Merck
All Photos(3)

Key Documents

SAB5702291

Sigma-Aldrich

Anti-GCN2 Antibody, clone 1G2Z4, Rabbit Monoclonal

Synonym(s):

GCN2, PVOD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

1G2Z4, monoclonal

form

liquid

mol wt

220 kDa

species reactivity

human, rat, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MAGGRGAPGRGRDEPPESYPQRQDHELQALEAIYGADFQDLRPDACGPVKEPPEINLVLYPQGLTGEEVYVKVDLRVKCPPTYPDVVPEIELKNAKGLSNESVNLLKSRLEELAKKHCGEVMIFELAYHVQSFLSEHNKPPPKSFHEEMLERRAQEEQQRLLEAKRKEEQEQREILHEIQRRKEEIKEEKKRKEMAKQERLEIASLSNQDHTSKKDPGGHRTAAILHGGSPDFVGNGKHRANSSGRSRRERQYSVCNSEDSPGSCEILYFNMGSPDQLMVHKGKCIGSDEQLGKLVYNAL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

This gene encodes a member of a family of kinases that phosphorylate the alpha subunit of eukaryotic translation initiation factor-2 (EIF2), resulting in the downregulaton of protein synthesis. The encoded protein responds to amino acid deprivation by binding uncharged transfer RNAs. It may also be activated by glucose deprivation and viral infection. Mutations in this gene have been found in individuals suffering from autosomal recessive pulmonary venoocclusive-disease-2. [provided by RefSeq, Mar 2014]

Immunogen

PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GCN2 (NP_001013725.2).

Application

WB, IHC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service