SAB5702056
Anti-SNRPA1 Antibody, clone 2D7M1, Rabbit Monoclonal
Synonym(s):
SNRPA1, Lea1, U2 small nuclear ribonucleoprotein A′
About This Item
biological source
rabbit
Quality Level
material
colorless
clone
2D7M1, monoclonal
form
liquid
mol wt
28 kDa
species reactivity
rat, human, rat
concentration
0.80 mg/mL
technique(s)
immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
immunogen sequence
QEAEKMFKGKRGAQLAKDIARRSKTFNPGAGLPTDKKKGGPSPGDVEAIKNAIANASTLAEVERLKGLLQSGQIPGRERRSGPTDDGEEEMEEDTVTNGS
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... SNRPA1(6627)
General description
Immunogen
Application
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service