SAB5201964
Monoclonal Anti-CD74-PE/Atto 594 antibody produced in mouse
clone 3D7
About This Item
Recommended Products
biological source
mouse
conjugate
PE/Atto 594 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
3D7, monoclonal
form
buffered aqueous solution
mol wt
33 kDa
species reactivity
human
concentration
1.0 mg/mL
technique(s)
flow cytometry: 1:100
western blot: 1:1000
isotype
IgG1
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CD74(972)
Related Categories
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service