SAB4702034
Monoclonal Anti-CD38 PE-Cyanine7 produced in mouse

Synonym(s):
ADP-ribosyl cyclase 1, ADPRC1, NAD(+) nucleosidase, T10, cADPr hydrolase 1
About This Item
conjugate
PE-Cy7 conjugate
Quality Level
antibody product type
primary antibodies
clone
HIT2, monoclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
flow cytometry: suitable
isotype
IgG1
UniProt accession no.
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... CD38(952)
General description
Specificity
Immunogen
Physical form
Preparation Note
Storage and Stability
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service