Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502769

Sigma-Aldrich

Anti-SHC antibody produced in rabbit

affinity isolated antibody

Synonym(s):

SH2 domain protein C1, SHC, SHC-transforming protein 1, SHCA, Src homology 2 domain-containing-transforming protein C1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 62 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SHC1(6464)

General description

Anti-SHC Antibody detects endogenous levels of total SHC protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Shc.

Immunogen Range: 315-364

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santosh Kumar et al.
Biochemical and biophysical research communications, 449(4), 496-501 (2014-05-23)
Hypercholesterolemia leads to a prothrombotic phenotype. Platelet hyperactivity associated with hypercholesterolemia has been attributed, in part, to oxidative stress. P66Shc is a well-known determinant of cellular and organismal oxidative stress. However, its role in platelet biology is not known. We
M Halon et al.
Free radical research, 48(11), 1363-1370 (2014-09-02)
Recently, iron and the adaptor protein "p66Shc" have been shown to play an important role in the development of amyotrophic lateral sclerosis (ALS) in rats. We hypothesized that changes in muscle p66Shc activity and iron metabolism would appear before visible
Doris Mangiaracina Benbrook et al.
Investigational new drugs, 32(3), 412-423 (2013-11-21)
SHetA2 is a small molecule flexible heteroarotinoid (Flex-Het) with promising cancer prevention and therapeutic activity. Extensive preclinical testing documented lack of SHetA2 toxicity at doses 25 to 150 fold above effective doses. Knowledge of the SHetA2 molecular target(s) that mediate(s)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service