Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300250

Sigma-Aldrich

Anti-phospho-SMAD1 (pSer465) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BSP1 antibody produced in rabbit, Anti-JV4-1 antibody produced in rabbit, Anti-JV41 antibody produced in rabbit, Anti-MADH1 antibody produced in rabbit, Anti-SMAD family member 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~55 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(I-S-S-V-SP)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer465)

Gene Information

human ... SMAD1(4086)

Immunogen

Peptide sequence around phosphorylation site of serine 465 (I-S-S-V-S(p)), according to the protein SMAD1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Transcriptional modulator activated by BMP (bone morphogenetic proteins) type 1 receptor kinase. SMAD1 is a receptor-regulated SMAD (R-SMAD).

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Wang et al.
Stem cell research, 39, 101497-101497 (2019-07-22)
Human embryonic stem cells (hESCs) have great potential for the repair of damaged articular cartilage. We developed a serum-free 14-day protocol for hESC differentiation into chondrocyte progenitors, which surprisingly lacked strong cartilage matrix production in in vitro tests. In order
Tomoko Watanabe et al.
Development (Cambridge, England), 145(20) (2018-10-07)
The pre-placodal ectoderm (PPE) is a specialized ectodermal region which gives rise to the sensory organs and other systems. The PPE is induced from the neural plate border during neurulation, but the molecular mechanism of PPE formation is not fully
Long Bai et al.
Endocrinology, 158(10), 3501-3511 (2017-10-05)
Bone morphogenetic protein 2 (BMP2) belongs to the transforming growth factor-β superfamily and plays a critical role in regulating ovarian follicle function. Currently, the role of BMP2 during cumulus expansion remains to be determined. The aim of this study was
José M Martín-Durán et al.
Nature ecology & evolution, 1(1), 5-5 (2017-08-16)
The mouth opening of bilaterian animals develops either separate from (deuterostomy) or connected to (protostomy) the embryonic blastopore, the site of endomesoderm internalization. Although this distinction preluded the classification of bilaterian animals in Deuterostomia and Protostomia, and has influenced major
Laurie M G de Kroon et al.
Cartilage, 8(4), 432-438 (2017-09-25)
Objective Previously, we demonstrated the importance of transforming growth factor-β (TGFβ)-activated SMAD2/3 signaling in chondrogenesis of bone marrow-derived mesenchymal stem cells (BMSCs). However, TGFβ also signals via the SMAD1/5/9 pathway, which is known to induce terminal differentiation of BMSCs. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service