Skip to Content
Merck
All Photos(2)

Key Documents

SAB1412193

Sigma-Aldrich

ANTI-T antibody produced in mouse

clone 5C8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC104817, T, TFT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5C8, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.63 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... T(6862)

General description

The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells. (provided by RefSeq)

Immunogen

T (NP_003172, 222 a.a. ~ 320 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ERSDHKEMMEEPGDSQQPGYSQWGWLLPGTSTLCPPANPHPQFGGALSLPSTHSCDRYPTLRSHRSSPYPSPYAHRNNSPTYSDNSPACLSMLQSHDNW

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana L Castillo-Carranza et al.
Journal of Alzheimer's disease : JAD, 40 Suppl 1, S97-S111 (2014-03-08)
Neurodegenerative disease is one of the greatest health crises in the world and as life expectancy rises, the number of people affected will continue to increase. The most common neurodegenerative disease, Alzheimer's disease, is a tauopathy, characterized by the presence
T B Smith et al.
Andrology, 2(5), 755-762 (2014-08-02)
We have shown previously that a network of mononuclear phagocytes (MPs) expressing macrophage and dendritic cell markers such as CD11c, F4/80 and CX3CR1, lines the base of the epididymal tubule. However, in the initial segment (IS) and only in that
Jessica Oenarto et al.
Archives of biochemistry and biophysics, 560, 59-72 (2014-07-09)
This study characterizes the expression of the osmolyte transporters betaine/γ-amino-n-butyric acid (GABA) transporter (BGT-1), the taurine transporter (TauT) and the sodium-dependent myo-inositol transporter (SMIT) in various rat brain cells in culture and in rat and human cerebral cortex in situ.
Akiko Ohtani et al.
Neuroscience research, 81-82, 11-20 (2014-04-05)
Serotonin (5-HT) regulates the development of cerebral cortex, but 5-HT receptors mediating the effects are poorly understood. We investigated roles of 5-HT2A receptor in dendritic growth cones using dissociation culture of rat cerebral cortex. Neurons at embryonic day 16 were
Olga Wiens et al.
PloS one, 9(7), e103821-e103821 (2014-08-01)
The invasion of Theileria sporozoites into bovine leukocytes is rapidly followed by the destruction of the surrounding host cell membrane, allowing the parasite to establish its niche within the host cell cytoplasm. Theileria infection induces host cell transformation, characterised by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service