Key Documents
OH30378545
Ohaus® Dissolved Oxygen Electrode
STDO21, cable L 5 m
About This Item
UNSPSC Code:
39121436
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30378545
dissolved oxygen measuring range
0.00-20.0 mg/L
cable L
5 m
General description
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Application
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Features and Benefits
Can be used in conjunction with all Starter bench and portable meters
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)