Skip to Content
Merck
All Photos(1)

Key Documents

MTCP8025LID

MTC Bio Reservoir Lid

capacity 25 mL, sterile, pack of 200 ea (40 x bags 5 ea)

Synonym(s):

pipetting tray

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122500
NACRES:
NB.12

material

polystyrene

sterility

sterile

packaging

pack of 200 ea (40 x bags 5 ea)

manufacturer/tradename

MTC Bio P8025-LID

capacity

25 mL

General description

Don’t leave valuable reagents in open reservoirs vulnerable to debris or airborne contaminants. Protective Lids inhibit evaporation and keep your solutions/reagents safe from dust or contamination.
Lids for 25mL solution reservoirs, sterile, 5 per bag

Legal Information

MTC is a trademark of MTC Bio

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service