MTCH680050
MTC™ Bio Halo™ Lightweight Single Channel Pipettor
capacity 5-50 μL
Synonym(s):
pipette
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.15
feature
adjustable
manual
no. of channels 1
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio H6800-50
capacity
5-50 μL
Related Categories
General description
As one of the lightest pipettors ever introduced, the Halo™ Pipettor dramatically reduces the risk of repetitive stress injury and the general shoulder, arm, and hand fatigue associated with daily pipetting. Reduced actuation forces andan ergonomic design make Halo™ Pipettors feel balanced and natural in hand. Featuring a four-digit micrometer display for precise volume selection and a click-based adjustment wheel with DriftGuard protection, the Halo™ Pipettor is perfect for applications that demand exceptional accuracy. With a recessed ejector button, and cushioned, shock-absorbing stop, the ergonomics of tip ejection have been re-imagined to relieve stress on the thumb and joints.
Halo™ Single Channel Pipettor, lightweight, 0.5-5mL
Features and Benefits
- 10-40% lighter than leading competitors
- 4 Digit micrometer with DriftGuard protection
- Exclusive 1250µL model for extended length tips
- User-enabled calibration port
- Compatible with universal tips
Legal Information
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)