Skip to Content
Merck
All Photos(1)

Key Documents

EHU051631

Sigma-Aldrich

MISSION® esiRNA

targeting human MEGF10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGTCAAAACGGAGCTGACTGCGACCACATTTCTGGGCAGTGTACTTGCCGCACTGGATTCATGGGACGGCACTGTGAGCAGAAGTGCCCTTCAGGAACATATGGCTATGGCTGTCGCCAGATATGTGATTGTCTGAACAACTCCACCTGCGACCACATCACTGGGACCTGTTACTGCAGCCCCGGATGGAAGGGAGCGAGATGTGATCAAGCTGGTGTTATCATAGTTGGAAATCTGAACAGCTTAAGCCGAACCAGTACTGCTCTCCCTGCTGATTCCTACCAGATCGGGGCCATTGCAGGCATCATCATTCTTGTCCTAGTTGTTCTCTTCCTACTGGCATTGTTCATTATTTATAGACACAAGCAGAAGGGAAAGGAATCAAGCATGCCAGCAGTTAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madhurima Saha et al.
Human molecular genetics, 28(14), 2365-2377 (2019-07-04)
MEGF10 myopathy is a rare inherited muscle disease that is named after the causative gene, MEGF10. The classic phenotype, early onset myopathy, areflexia, respiratory distress and dysphagia, is severe and immediately life-threatening. There are no disease-modifying therapies. We performed a
Jessica Charlet et al.
Molecular carcinogenesis, 56(4), 1290-1301 (2016-11-20)
Neuroblastoma is a childhood cancer in which many children still have poor outcomes, emphasising the need to better understand its pathogenesis. Despite recent genome-wide mutation analyses, many primary neuroblastomas do not contain recognizable driver mutations, implicating alternate molecular pathologies such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service