Key Documents
BR701377
BRAND® sealing film for microplates
new
suitable for PCR, storage, aluminum film, DMSO/solvent resistant, operating temperature (-80)—120 °C, pack of 100 ea
Synonym(s):
701377, BRAND film, PCR sealing film, aluminum film, cover, plate film, seal, sealing film, storage film
About This Item
UNSPSC Code:
41122410
material
aluminum film
sterility
non-sterile
feature
DNA free
RNase and DNase free
packaging
pack of 100 ea
manufacturer/tradename
BRAND 701377
parameter
(-80)—120 °C operating temperature
suitability
suitable for PCR
General description
This film is ideal for PCR, ELISA and sample storage. Due to its adhesive, it can be used down to -80°C, is DMSO-resistant, and resistant to common solvents. It can be removed without leaving any residue so that work can continue unhindered after transport or storage.
Features and Benefits
  • Temperature stability to 120°C and down to -80°C
  • Tight seal for minimizing evaporation
  • DMSO- and solvent-resistant
  • Removable without leaving residue
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)