BAF409410000
Bel-Art® Conway Diffusion Cell
O.D. 83 mm, pack of 3 ea
Synonym(s):
Conway diffuser, Obrink modification, diffusion analysis of blood, microdiffusion analysis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41123100
NACRES:
NB.22
Recommended Products
packaging
pack of 3 ea
manufacturer/tradename
Bel-Art F40941-0000
O.D.
83 mm
center depth × diam.
4 mm × 35 mm
chamber depth × diam.
10 mm × 60 mm
General description
A knob on the cover is used to raise it, and the circular moat of the cell receives the cover, sealing off the outermost chamber. Sterilizable at 121ºC (250ºF). 3 per bag
Assay CO2, Ammonia or alcohol levels in blood or serum using the diffusion method
Assay CO2, Ammonia or alcohol levels in blood or serum using the diffusion method
Features and Benefits
- Obrink modification of the Conway Diffusion Cell
- Transparent polypropylene cover
- Autoclavable
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service