Skip to Content
Merck
All Photos(1)

Key Documents

AV31924

Sigma-Aldrich

Anti-TBX21 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-T-box 21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

58 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TBX21(30009)

General description

Rabbit polyclonal anti-TBX21 (AB1) antibody reacts with human, bovine, canine, and mouse T-box transcription factor 21 transcription factors.
T-box genes encode transcription factors that regulate developmental processes. T-box transcription factor 21 (TBX21) is the human ortholog of mouse Tbx21/Tbet, a lineage commitment regulator for the differentiation of interferon-gamma (IFNG) producting CD4 T helper (Th1) 1 cells.
TBX21 is a transcription factor that is involved in the development of T lymphocytes. Functional TBX21 variations have been associated with aspirin-induced asthma and clinical outcomes for inhaled corticosteroid therapy in asthma patients.

Immunogen

Synthetic peptide directed towards the N terminal region of human TBX21

Application

Rabbit Anti-TBX21 (AB1) antibody can be used for western blot assays at a concentration of 2.5μg/ml.
Rabbit polyclonal anti-TBX21 (AB1) antibody is used to tag T-box 21 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of T-box transcription factor 21 in the differentiation of interferon-gamma (IFNG) producting CD4 T helper 1 (Th1) cells.

Biochem/physiol Actions

TBX21 is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Expression of the human TBX21 correlates w

Sequence

Synthetic peptide located within the following region: FYPEPGAQDADERRGGGSLGSPYPGGALVPAPPSRFLGAYAYPPRPQAAG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitsuteru Akahoshi et al.
Human genetics, 117(1), 16-26 (2005-04-05)
Asthma is a phenotypically heterogeneous disorder with many etiologic factors and clinical characteristics. T-bet, a Th1-specific transcription factor of T-box family, has been found to control interferon-gamma (IFN-gamma) expression in T cells. Mice lacking the T-bet gene (tbx21) demonstrate multiple
Kelan G Tantisira et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(52), 18099-18104 (2004-12-18)
TBX21 encodes for the transcription factor T-bet (T-box expressed in T cells), which influences naive T lymphocyte development and has been implicated in asthma pathogenesis. Specifically, the T-bet knockout mouse spontaneously develops airway hyperresponsiveness and other changes consistent with asthma.
Judy T Tellam et al.
PLoS pathogens, 10(10), e1004423-e1004423 (2014-10-10)
Recent studies have shown that virally encoded mRNA sequences of genome maintenance proteins from herpesviruses contain clusters of unusual structural elements, G-quadruplexes, which modulate viral protein synthesis. Destabilization of these G-quadruplexes can override the inhibitory effect on self-synthesis of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service