Skip to Content
Merck
All Photos(1)

Key Documents

936871

Sigma-Aldrich

AdBrettPhos Pd G3 ChemBeads

Synonym(s):

Di-Ad-BrettPhos-G3-Palladacycle, [2-(Di-1-adamantylphosphino)-2′,4′,6′-triisopropyl-3,6-dimethoxybiphenyl][2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H74NO5PPdS
CAS Number:
Molecular Weight:
1010.65
UNSPSC Code:
12352100
NACRES:
NA.21

form

solid

Quality Level

composition

, ~5 wt. % (loading of catalyst)

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

COC1=CC=C(OC)C(C2=C(C(C)C)C=C(C(C)C)C=C2C(C)C)=C1P([C@]34C[C@H]5C[C@H](C[C@H](C5)C4)C3)[C@@]6(C[C@H](C7)C8)CC7C[C@H]8C6.NC9=CC=CC=C9C%10=CC=CC=C%10[Pd]OS(C)(=O)=O

InChI

InChI:1S/C43H61O2P.C12H10N.CH4O3S.Pd/c1-25(2)34-17-35(26(3)4)39(36(18-34)27(5)6)40-37(44-7)9-10-38(45-8)41(40)46(42-19-28-11-29(20-42)13-30(12-28)21-42)43-22-31-14-32(23-43)16-33(15-31)24-43;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h9-10,17-18,25-33H,11-16,19-24H2,1-8H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

OZFYYHNERORONM-UHFFFAOYSA-M

General description

Buchwald 3rd Generation Pd Precatalyst adorned by the bulk di-adamntyl BrettPhos ligand. This complex has been used in the monoarylation of ammonia, displaying aptitude for otherwise difficult 5-member heterocyclic substrates.

Application

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

For larger scale uses, product also available in powdered form (776106)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service