936774
PPh3 Pd G2 ChemBeads
Synonym(s):
Chloro(triphenylphosphine) [2-(2′-amino-1,1′-biphenyl)]palladium(II), Triphenylphosphine Pd G2
About This Item
Recommended Products
form
solid
Quality Level
composition
, ~5 wt. % (loading of catalyst)
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
SMILES string
Nc1ccccc1-c2ccccc2[Pd]Cl.c3ccc(cc3)P(c4ccccc4)c5ccccc5
InChI
1S/C18H15P.C12H10N.ClH.Pd/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h1-15H;1-6,8-9H,13H2;1H;/q;;;+1/p-1
InChI key
RNURSGDBDHOTOE-UHFFFAOYSA-M
Related Categories
General description
Application
For larger scale uses, product also available in powdered form (752762)
Other Notes
Versatile Methods to Dispense Sub-Milligram Quantities of Solids using Chemical Coated Beads for High-Throughput Experimentation
ChemBead Enabled High-Throughput Cross-Electrophile Coupling Reveals a New Complementary Ligand
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service