Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB197F

Sigma-Aldrich

Milli-Mark® Anti-CD43-FITC Antibody, clone DF-T1

clone DF-T1, Milli-Mark®, from mouse

Synonym(s):

Leukosialin, Leukocyte sialoglycoprotein, Sialophorin, Galactoglycoprotein, GALGP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

DF-T1, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SPN(6693)

General description

CD43 is a human heavily 0-glycosylated transmembrane protein, which is expressed by T cells and myeloid lineage cells (Borche, 1987; Stross, 1989). Reactivity with plasma cells has been observed (Beschomer, 1999). The molecule is known to be defective in the Wiskott-Aldrich syndrome and may participate in T-cell activation (Stross, 1989).
Immunoblotting studies have revealed molecular mass heterogeneity displayed by the antigen extracted from white blood cells of different lineage. Thus, two major bands of 140 and 130 kDa are seen for the promyelocytic cell line HL60, 110 kDa for the myeloblast cell line KG1 and a major 115 kDa band for peripheral blood lymphocytes (Stross, 1989).

Specificity

Antibody recognizes Human CD43 antigen.

Immunogen

Myeloblast cell line KG1

Application

Detect CD43 using this Milli-Mark Anti-CD43-FITC Antibody, clone DF-T1 validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using human PBMCs

Target description

110 kDa (Stross, 1989)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintained refrigerated at 2-8°C from date of receipt. Protected from light in undiluted aliquots for 6 months from date of receipt.

Analysis Note

Control
PBMCs

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service