Skip to Content
Merck
All Photos(1)

Key Documents

870742P

Avanti

4ME 16:0 Coenzyme A

Avanti Research - A Croda Brand 870742P, powder

Synonym(s):

phytanoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H83N10O17P3S
CAS Number:
Molecular Weight:
1113.14
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (870742P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870742P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CC(C)CCCC(C)CCCC(C)CCCC(C)C)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C41H74N7O21P3S/c1-25(2)11-8-12-26(3)13-9-14-27(4)15-10-16-28(5)21-31(50)73-20-19-43-30(49)17-18-44-38(53)35(52)40(6,7)41(54,55)66-67-72(61,62)69-68-71(59,60)63-22-29-34(65-70(56,57)58)33(51)39(64-29)48-24-47-32-36(42)45-23-46-37(32)48/h23-29,33-35,39,51-52,54-55H,8-22H2,1-7H3,(H,43,49)(H,44,53)(H,59,60)(H,61,62)(H2,42,45,46)(H2,56,57,58)/t26-,27+,28+,29+,33-,34?,35-,39-/m1/s1

InChI key

LKWLNEKCMODJGW-WBEGNQIBSA-N

General description

4ME 16:0 Coenzyme A, also known as phytanoyl coenzyme A, is a coenzyme A derivative of phytanic acid. Phytanoyl coenzyme A is a metabolic intermediate of phytol degradation pathway. It is converted to 2-hydroxyphytanoyl-CoA by the enzyme phytanoyl-CoA 2-hydroxylase (PAHX). Accumulation of phytanic acid due to the deficiency of PAHX causes Refsum′s disease.

Packaging

5 mL Amber Glass Screw Cap Vial (870742P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N J Kershaw et al.
Bioorganic & medicinal chemistry letters, 11(18), 2545-2548 (2001-09-11)
Phytanoyl-CoA 2-hydroxylase (PAHX), an iron(II) and 2-oxoglutarate-dependent oxygenase, catalyses an essential step in the mammalian metabolism of beta-methylated fatty acids. Phytanoyl-CoA was synthesised and used to develop in vitro assays for PAHX. The product of the reaction was confirmed as
Phytanoyl-coenzyme A hydroxylase deficiency -- the enzyme defect in Refsum's disease.
G A Jansen et al.
The New England journal of medicine, 337(2), 133-134 (1997-07-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service