Skip to Content
Merck
All Photos(1)

Key Documents

Z303976

Quickfit® jacketed reaction flasks

capacity 2,000 mL

Synonym(s):

JRV2L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804

material

borosilicate glass

sterility

non-sterile

packaging

pack of 1

manufacturer/tradename

Quickfit JRV 2L

availability

not available in USA and Canada

capacity

2,000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Water jacket for heating or cooling flask contents. For lids and clamp please order separately.

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service