Skip to Content
Merck
All Photos(1)

Key Documents

GF96200344

Platinum-iridium alloy, Pt90Ir10

1m insulated wire, 0.025mm conductor diameter, 0.005mm insulation thickness, ptfe (polytetrafluoroethylene) insulation, 99.999%

Synonym(s):

PT025825

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

description

ptfe (polytetrafluoroethylene) insulation

Assay

99.999%

form

wire (insulated)

manufacturer/tradename

Goodfellow 962-003-44

L

1 m

conductor diam.

0.025 mm

insulation thickness

0.005 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service