Skip to Content
Merck
All Photos(2)

Key Documents

GF88318266

Zinc

wire reel, 50m, diameter 2.0mm, extruded, 99.9%

Synonym(s):

Zinc, ZN005135

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 487 °C)

Assay

≥99.9%

form

wire

manufacturer/tradename

Goodfellow 883-182-66

resistivity

5.8 μΩ-cm, 20°C

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johan Hattne et al.
Nature methods, 11(5), 545-548 (2014-03-19)
X-ray free-electron laser (XFEL) sources enable the use of crystallography to solve three-dimensional macromolecular structures under native conditions and without radiation damage. Results to date, however, have been limited by the challenge of deriving accurate Bragg intensities from a heterogeneous
Kohei Takeshita et al.
Nature structural & molecular biology, 21(4), 352-357 (2014-03-04)
The voltage-gated proton channel Hv1 (or VSOP) has a voltage-sensor domain (VSD) with dual roles of voltage sensing and proton permeation. Its gating is sensitive to pH and Zn(2+). Here we present a crystal structure of mouse Hv1 in the
Priscilla K Walker et al.
Health physics, 106(4), 523-527 (2014-02-25)
Beam shaping in scattered and uniform scanned proton beam therapy (PBT) is made commonly by brass apertures. Due to proton interactions, these devices become radioactive and could pose safety issues and radiation hazards. Nearly 2,000 patient-specific devices per year are
Shinya Tashiro et al.
Biochemistry, 53(14), 2218-2220 (2014-04-05)
Mutations of DJ-1 cause familial Parkinson's disease (PD), although the role of DJ-1 in PD remains unresolved. Very recent reports have shown that DJ-1 interacts with copper ions. This evidence opens new avenues to understanding the function of DJ-1 and
J Granell
The Journal of sports medicine and physical fitness, 54(2), 232-237 (2014-02-11)
The role of zinc and copper has been shown essential in the scope of physical exercise. The outcomes of the studies about changes in the concentrations of these elements in blood, in physical effort situations, are sometimes discordant and seem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service