GF82573836
Aluminum-magnesium alloy, Al95Mg5
1000mm tube, 1.73mm outside diameter, 0.075mm wall thickness, 1.58mm inside diameter, as drawn
Synonym(s):
AL027110
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352311
NACRES:
NA.23
form
tubes
manufacturer/tradename
Goodfellow 825-738-36
L × wall thickness
1000 mm × 0.075 mm
O.D. × I.D.
1.73 mm × 1.58 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service