GF22451408
Carbon
rod, 150mm, diameter 6.35mm, graphite, 100%
Synonym(s):
Activated charcoal, C 007921, Charcoal activated
About This Item
Recommended Products
vapor pressure
<0.1 mmHg ( 20 °C)
Assay
100%
form
rod
autoignition temp.
842 °F
manufacturer/tradename
Goodfellow 224-514-08
resistivity
1375 μΩ-cm, 20°C (graphite)
size diam.
150 mm × 6.35 mm
mp
3550 °C (lit.)
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service