Skip to Content
Merck
All Photos(1)

Key Documents

DWK90K16100R

Kimble® Tube Clt W/O Cap 16X100Mm, M/Cs

M/Cs

Synonym(s):

90K16100R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121706
NACRES:
NB.17

feature

cap: no

packaging

pkg of 1 packs

manufacturer/tradename

Kimble® 90K16100R

diam. × H

16 mm × 100 mm

Application

Bacteriology, RIA, coagulation and other laboratory procedures

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service