CLS5840200
Pyrex® volumetric flask, phosphoric acid
capacity 200 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass, Corning® 5840
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
feature
class B
packaging
case of 12 ea
case of 6 ea
manufacturer/tradename
Corning 5840-200
tol. measuring range
0.18 mL
capacity
200 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For use in determining phosphoric acid in mixed fertilizer. Of heavy construction throughout and with a wide neck.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service