Skip to Content
Merck
All Photos(1)

Key Documents

940895

Sigma-Aldrich

Iron(III) phosphate dihydrate

new
greener alternative

≥99.99% trace metals basis

Synonym(s):

Ferric orthophosphate, Ferric phosphate, Ferric phosphate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FePO4 · 2H2O
CAS Number:
Molecular Weight:
186.85
MDL number:
form:
powder or crystals
solubility:
water: soluble

grade

(Titration with Na2S2O3)

Quality Level

Assay

≥99.99% trace metals basis (≤ 100 ppm trace metals)
≥99.99% trace metals basis

form

powder or crystals

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

solubility

water: soluble

anion traces

chloride (Cl-): ≤20 ppm
sulfate (SO42-): ≤50 ppm

cation traces

Al: ≤10 ppm
Cd: ≤10 ppm
Cr: ≤10 ppm
K: ≤10 ppm
Mg: ≤10 ppm
Mn: ≤10 ppm
Na: ≤20 ppm
Ni: ≤10 ppm
Pb: ≤10 ppm
Zn: ≤10 ppm

greener alternative category

SMILES string

O.O.[Fe+3].[O-]P([O-])([O-])=O

InChI

1S/Fe.H3O4P.2H2O/c;1-5(2,3)4;;/h;(H3,1,2,3,4);2*1H2/q+3;;;/p-3

InChI key

BMTOKWDUYJKSCN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Iron(III) phosphate dihydrate, with a purity of ≥99.99% trace metals, is a high-purity chemical. It is a hydrated form of iron(III) phosphate, appearing typically as a solid with a slightly off-white to light yellow color. It comes with a purity ≥99.99% , and trace metal content ≤ 100 ppm. Its high purity makes it suitable for sensitive applications where trace metal contamination must be minimized.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product belongs to an Enabling category. Iron phophate dihydrate precursor enables greener synthesis of high-performance LiFePO4/C cathodes, demonstrating sustainable production for advanced lithium-ion batteries, offering a promising solution in greener energy storage technology. Click here for more information.

Application

Iron(III) phosphate dihydrate with a purity of ≥99.99% trace metals can be used in various research applications. It is suitable to use as a precursor in the production of lithium iron phosphate (LiFePO4) for use in lithium-ion batteries (LIBs) particularly in the cathode material. It can be used as an efficient catalyst for the chemo selective acetylation of alcohols and phenols under solvent free condition at room temperature and with high yields. Apart from the product also widely used in waste water purification and ferroelectric applications. [,]

Features and Benefits

With 99.99% purity, the product contains minimal trace metal impurities, making it ideal for use in sensitive processes where contamination must be strictly controlled. The high purity ensures consistent performance, which is essential in research and production environments that require reproducibility and precision.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service