Skip to Content
Merck
All Photos(1)

Key Documents

940194

Sigma-Aldrich

Lithium peroxide

new

99.9% trace metals basis, battery grade

Synonym(s):

Dilithium peroxide, Lithium(I) peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Li2O2
CAS Number:
Molecular Weight:
45.88
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:

grade

battery grade

Quality Level

Assay

99.9% trace metals basis

form

powder

solubility

acetic acid: soluble
alcohol: insoluble
water: soluble

density

2.32 g/cm3 (lit.)

application(s)

battery manufacturing

SMILES string

[Li+].[Li+].[O-][O-]

InChI

1S/2Li.O2/c;;1-2/q2*+1;-2

InChI key

HPGPEWYJWRWDTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our battery grade lithium peroxide product is a high-purity material. It meets a minimum purity of 99.9%, as determined by trace metals analysis, and is processed to a particle size after passing through a sieve with 100-mesh openings, roughly 150 microns in diameter.
Lithium peroxide is an inorganic compound with the composition Li2O2 that takes the form of a white powder. Chemically, lithium peroxide is strongly basic and a powerful oxidizing agent. At room temperature, it is stable in air, showing neither hygroscopic properties nor a tendency to absorb carbon dioxide. At elevated temperatures in air, Li2O2 and CO2 establish a reversible chemical equilibrium with lithium carbonate and oxygen. At even higher temperatures, ~340-450 °C, lithium peroxide decomposes to lithium oxide (Li2O) and oxygen gas.

Application

The primary use of our high-purity lithium peroxide is as a synthetic precursor in the production of high-purity lithium oxide and other lithium metal oxides. In battery research, lithium peroxide is a key precursor for lithium metal oxides, especially lithium-rich oxides with the disordered rock salt structure, which serve as cathode materials in lithium-ion batteries. Often, lithium peroxide is used in mechanochemical reactions, in which lithium peroxide is ball-milled with one or more metal oxides, followed by sintering at elevated temperatures. In this application, the trace metals purity of our lithium peroxide is critical, as the purity of the precursors directly influences the purity of the final product. Additionally, because of its high oxygen content by weight and its facile redox chemistry, lithium peroxide is also used in air purifiers to remove carbon dioxide, most notably on spacecraft, when weight is important. The reversible lithium peroxide reaction with carbon dioxide to form lithium carbonate and oxygen gas is also the chemical basis for lithium-air batteries.

related product

Product No.
Description
Pricing

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service