Skip to Content
Merck
All Photos(1)

Key Documents

938645

Sigma-Aldrich

N6-[[(1α,8α,9α)-Bicyclo[6.1.0]non-4-yn-9-ylmethoxy]carbonyl]-L-lysine

new

≥95%

Synonym(s):

L-Lysine, N6-[[(1α,8α,9α)-bicyclo[6.1.0]non-4-yn-9-ylmethoxy]carbonyl]-, Nε-(((1R,8S)-Bicyclo[6.1.0]non-4-yn-9-ylmethoxy)carbonyl)-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26N2O4
CAS Number:
Molecular Weight:
322.40
MDL number:

Quality Level

Assay

≥95%

form

powder or crystals

color

white to off-white

storage temp.

2-8°C

SMILES string

C(OC(NCCCC[C@@H](C(O)=O)N)=O)[C@H]1[C@]2([C@@]1(CCC#CCC2)[H])[H]

InChI

InChI=1S/C17H26N2O4/c18-15(16(20)21)9-5-6-10-19-17(22)23-11-14-12-7-3-1-2-4-8-13(12)14/h12-15H,3-11,18H2,(H,19,22)(H,20,21)/t12-,13+,14+,15-/m0/s1

InChI key

QLDVOCPEKYLEOT-YJNKXOJESA-N

Application

N6-[[(1α,8α,9α)-Bicyclo[6.1.0]non-4-yn-9-ylmethoxy]carbonyl]-L-lysine is particularly suited for the labeling and crosslinking of proteins on the surface of live mammalian cells. Its high reactivity and biocompatibility allow for the precise modification of target proteins with minimal off-target effects and without compromising cell viability. This capability is crucial for studying protein dynamics, interactions, and functions in real time, providing insights into cellular processes that were previously unattainable with traditional biochemical methods.

Features and Benefits

N6-[[(1α,8α,9α)-Bicyclo[6.1.0]non-4-yn-9-ylmethoxy]carbonyl]-L-lysine unique structure, offers unparalleled reactivity towards azide-functionalized biomolecules facilitating rapid and specific conjugation reactions without the need for cytotoxic copper catalysts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service