Skip to Content
Merck
All Photos(1)

Key Documents

938394

Sigma-Aldrich

Strontium

new

beads, ≤2.0 mm particle size, 99.99% trace metals basis

Synonym(s):

Elemental Barium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sr
CAS Number:
Molecular Weight:
87.62
MDL number:
UNSPSC Code:
12141504

Quality Level

Assay

99.99% trace metals basis

form

beads

resistivity

23 μΩ-cm, 20°C

particle size

≤2.0 mm

bp

1384 °C (lit.)

mp

757 °C (lit.)

density

2.6 g/mL at 25 °C (lit.)

SMILES string

[Sr]

InChI

1S/Sr

InChI key

CIOAGBVUUVVLOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Strontium is a divalent alkaline earth metal with a silvery luster. It is an active reducing agent and readily reacts with oxygen, sulfur, and halogen. It is widely used in the preparation of Sr-based optical materials for piezoelectric, electro-optic devices, and bioactive glasses for bone regeneration.

Application

For the synthesis of biomaterials and superconducting materials, the raw materials must be pure. Therefore, in order to achieve the desired performances, it is crucial to reduce undesired metals or interfering elements. For the synthesis of premium new materials, our 99.99% trace metal-based strontium beads are the best option.
Elemental Strontium has wide applications. For example, Superconducting Strontium Germanide (SrGe3) Containing Ge2 Dumbbells is synthesized at high pressure and high temperture using metallic Stronium. Strontium is also used in low temperature solution process to synthesize strontium doped oxide with high carrier mobility, enhanced electrical stability for thin film transition.
In addition, Strontium is applied in the solid state synthesis of a Zintl phase, SrSn3Sb4, with an anionic channel framework of 30-membered rings. Interestingly, Strontium is used to synthesis the crack free Strontium-potassium-niobate (Sr2KNb5O15, SKN) thin films by using chemical solution deposition method. SKN is of great interest for pyroelectric sensors and various electro-optic devices.
Further more, Strontium (Sr) has also been investigated as a bioactive element capable of stimulating bone tissue regeneration.According to the literature, bioactive-glass incorporated with Sr has recently received a lot of attention for various orthopaedic applications that combine the effect of BG surface bioactivity with the promotion of bone growth due to Sr-controlled release.[5]
Used to make Strontium-potassium-niobate (Sr2KNb5O15, SKN). SKN is of great interest for pyroelectric sensors and various electro-optic devices.2

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service