Skip to Content
Merck

8325-OP

Millipore

Tris Buffered Saline

20X Liquid Concentrate, OmniPur® Grade

Synonym(s):

TBS

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Tris Buffered Saline, OmniPur® Grade, 20X Ready Pack Powder

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

color

clear

shipped in

ambient

General description

Each pack makes 1 L of 20X buffer solution. The unit contains two packs. 1X Solution contains 137 mM sodium chloride, 2.7 mM potassium chloride and 2 5mM Tris/Tris-HCl.

Specifications:
Molarity of Tris 450-550 mM
pH at 25°C: 7.25-7.55





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

8325-2PACK:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service