EMU048061
MISSION® esiRNA
targeting mouse Ddx17
description
Powered by Eupheria Biotech
Quality Level
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
TGATGGATGTGCTGATGGATCAGCACTTTACAGAACCCACTCCCATTCAGTGCCAGGGATTTCCTTTGGCTCTTAGTGGCAGGGATATGGTTGGCATTGCACAGACTGGCTCTGGGAAGACATTGGCGTATTTGCTGCCTGCGATTGTTCATATAAACCACCAGCCATACTTGGAAAGAGGAGATGGTCCAATTTGTCTAGTGCTGGCTCCTACCAGAGAGCTTGCGCAGCAGGTGCAGCAGGTGGCTGACGATTATGGAAAATGCTCCAGGTTGAAGAGTACGTGCATTTACGGAGGTGCTCCTAAAGGTCCCCAAATTCGAGACTTGGAAAGAGGTGTTGAGATTTGCATAGCCACTCCTGGGCGCCTAATAGATTTCCTGGAGTCAGGAAAGACAAACCTTCGCCGATGTAC
Ensembl | mouse accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... DDX17(67040), Ddx17(67040)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.