Skip to Content
Merck

SAE0220

Mucinase SmE

mass spec suitable, from Serratia marcescens, recombinant, liquid

Synonym(s):

SmEnhancin

Sign Into View Organizational & Contract Pricing

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

recombinant

expressed in E. coli

Quality Level

form

liquid

shelf life

2 yr (Retest)

mol wt

98 kDa

suitability

suitable for mass spectrometry

shipped in

dry ice

storage temp.

-10 to -25°C

General description

Mucinase SmE is a mucin-selective protease that has been engineered to have high activity and and tolerance for site ocupancy and glycoform complexity. Mucinase SmE provides a complementary cleavage profile to Mucinase StcE (Cat No SAE0202). Contrary to StcE digestion that requires an additional tryptic digest prior to MS peptide analysis, when using Mucinase SmE analysis can be done with SmE alone. It is recommended to perform both options to obtain maximal coverage.

Application

Mucinase SmE is useful for glycoproteomics experimentation. It is designed for digestion of densely glycosylated mucin proteins for mass spec based analyses.

Features and Benefits

  • Mucin-selective Protease
  • Cleaves N-terminally to O-link glycosylated Ser/Thr in mucin domains
  • Complimentary cleavage profile to Mucinase StcE
  • Accommodates complex, dense and sialylated glycoforms
  • Recombinant with His-Tag®

Preparation Note

Thaw the Mucinase SmE by brief incubation at 37°C.

The protein solution is stable for 2 weeks at 2-8 °C.

For longer storage, aliquot the protein solution and store at -20 °C.

The enzyme solution should be protected from direct light.

Legal Information

HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service