1.03736
StripScan Reference Card
for use with pH-indicator strips
product line
MQuant®
compatibility
for use with MQuant® StripScan App
for use with pH-indicator strips
detection method
colorimetric
storage temp.
15-25°C
General description
for the measurement of MQuant® pH-indicator strips pH 0 – 14, Cat. No. 109535 MQuant
Legal Information
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service