SAB2109156
Anti-IL6ST antibody produced in rabbit
affinity isolated antibody
Synonym(s):
CD130, CDw130, GP130, GP130-RAPS, IL6R-beta
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
35 kDa
species reactivity (predicted by homology)
bovine, horse, canine, human, rat, rabbit, guinea pig
concentration
0.5 mg/mL
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 4-8 μg/mL
western blot: 1 μg/mL
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... IL6ST(57396)
General description
Sequence
Physical form
Disclaimer
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.