939897
Lithium cobalt oxide

lithium-niobate coated LCO cathode active material
Synonym(s):
Lithium cobalt(III) oxide, LCO, LiNbO3-coated LCO, Lithium cobaltite
grade
battery grade
Quality Level
Assay
>99%
form
powder
mol wt
97.87 g/mol
composition
LiCoO2
density
4.82 g/cm3 (lit.)
application(s)
battery manufacturing
SMILES string
[Li+].[O-][Co]=O
InChI
1S/Co.Li.2O/q;+1;;-1
InChI key
BFZPBUKRYWOWDV-UHFFFAOYSA-N
General description
Application
Features and Benefits
- LCO classic model system for fundamental research
- LiNbO3 coating safeguards cathode and electrolyte, enhancing coulombic
- LiNbO3 insertion layer enhances rate capability
Still not finding the right product?
Explore all of our products under Lithium cobalt oxide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1B - Skin Sens. 1
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.